Learn More
Chemglass Life Sciences Extraction Apparatus, Jumbo, Size C
SDP
Supplier:  Chemglass Life Sciences CG137303
Encompass_Preferred
Extraction Apparatus, Jumbo, Size C. Jumbo Soxhlet extraction apparatus equipped with a Duran flange at the top of the extractor. Flange is formed with a groove in the flat portion of the mating surface to accept a PTFE covered silicone o-ring to permit grease free assembly. The adapter between the condenser and extractor has a top outer joint and a Duran flange which attaches to the extractor body with a CG-141 stainless steel clamp. Complete apparatus consists of a condenser, extractor, round bottom flask, adapter, PTFE covered o-ring and clamp. PTFE sleeves and heating mantles must be ordered separately.Extractor Capacity (mL) 5,000 Extractor I.D. (mm) 140 Extractor Flange Size 150 Extractor Lower Joint 55/50 Condenser Length (mm) 750 Paper Thimble Size (mm) 125 x 310 Flask Capacity (mL) 12,000
Catalog No. CG137303
Explore available promotions