Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE Microporous Cleanroom boot. Non-skid sole. 19" upper. Individually packaged in pairs. 50 pr/case with overlay bag. Size LGXL
Supplier: Lakeland Industries CTL903CMPLGXL
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold on each boot. Bound seams provide a superior barrier to prevent breakthrough and strike through. Boots are constructed with a PFA free, fluid-repellant, low-linting, microporous fabric and non-skid sole. Encased elastic and 19" upper. Individually packaged in pairs, 50 pr /case. Size LGXL. Nonsterile.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.