Learn More
Drg International Inc CHROMagar™ Listeria, dehydrated culture media, 5000 mL reconstituted
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Drg International Inc LM852
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
CHROMagar™chromogenic plates are easy to prepare - just add water to the dehydrated media, bring to a boil, let it cool down and pour! Use the entire pack, or if there is a need for a smaller number of plates, just a portion. CHROMagar™ products are sold in dehydrated form. DRG offers the 5L size (after reconstitution). If kept under appropriate storage temperatures, CHROMagar™ powder products have a long shelf life (2-3 years). This flexibility is essential to avoid the costly waste resulting from unused plates. This product requires a refrigerated supplement to use properly; these are shipped free of charge along with the product. Supplements contain compounds that cannot be heated, therefore the compounds would degrade in the base if stored together for a long time.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.