Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloride Standard 500 ppm Cl (0.0141 Normal), Reagents
Chloride Standard 500 ppm Cl (0.0141 Normal), Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
Specifications
Chemical Name or Material | Chloride Standard 500 ppm Cl (0.0141 Normal) |
---|---|
CAS | 7732-18-5, 7647-14-5 |
Packaging | Poly Bottle |
Color | Colorless |
For Use With (Equipment) | Water Analyzer |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-335-631
|
Reagents Holdings Llc
CC1554001A |
1 L |
Each for $146.20
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-335-630
|
Reagents Holdings Llc
CC155400500A |
500 mL |
Each for $104.72
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Specifications
Chloride Standard 500 ppm Cl (0.0141 Normal) | |
Poly Bottle | |
Water Analyzer | |
Reagent | |
100°C | |
Water Analysis | |
Liquid |
7732-18-5, 7647-14-5 | |
Colorless | |
NaCl/H2O | |
6.7 to 7.3 | |
1.001 | |
Completely miscible |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title