Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Chicken Lysozyme ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of chicken lysozyme.
Supplier: Novus Biologicals™ NBP2600881KIT
Description
The Chicken Lysozyme ELISA Kit (Colorimetric) measures chicken Lysozyme in serum, plasma and egg white.Specifications
15.625 to 4000ng/mL | |
ELISA Kit (Colorimetric) | |
For detection of chicken lysozyme | |
ELISA | |
LYZ | |
1 Kit | |
Cardiovascular Biology | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Lysozyme |
14.876ng/mL | |
Egg White, Plasma, Serum | |
Chicken | |
4069 | |
Adhesive Strip (For 96 wells), Assay plate, Biotin-conjugate (100 x concentrate), Biotin-conjugate Diluent, HRP-avidin Diluent, HRP-avidin (100 x concentrate), TMB Substrate, Wash Buffer (25 x concentrate), Stop Solution | |
RUO | |
50 to 100 μL | |
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction