Learn More
MilliporeSigma™ Chemicon™ Human Collagen Type I
Initially produced as procollagen in cells
Supplier: MilliporeSigma™ CC050
Description
- COL1A1 is gene responsible for production of alpha1(I) chain of type I collagen
- Consists of two pro-alpha1(I) protein strands combined with a pro-alpha2(I) procollagen strand that form a triple-stranded rope-like structure
Specifications
1 mg/mL | |
100 μg | |
Collagen, Human Type I | |
Liquid |
Cell Culture | |
Human placenta, negative for HBsAG and HIV antibodies | |
Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
>90% collagen type I |