Learn More
MALIX LLC The Chembio DPP HIV-Syphilis test is an FDA-approved, dual rapid test for detecting HIV 1/2 and syphilis antibodies. Suitable for fingerstick, venous whole blood, or plasma. Quick results for timely diagnosis.
Supplier: MALIX LLC DPPHIVSYPH20PK
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Chembio DPP HIV-Syphilis
The Chembio DPP HIV-Syphilis test is an FDA-approved dual rapid test designed for the detection of antibodies to HIV 1/2 and Treponema pallidum (syphilis). This test can be performed using fingerstick whole blood, venous whole blood, or plasma specimens, offering flexibility in sample collection. It is intended for use in clinical settings where rapid and reliable results are essential for timely diagnosis and treatment. 5PK/20Kits
Key Features:
FDA Approved: Ensures high standards of accuracy and reliability.
Dual Detection: Simultaneously detects antibodies to HIV 1/2 and Treponema pallidum.
Specimen Collection: Suitable for fingerstick whole blood, venous whole blood, or plasma specimens.
Rapid Results: Provides quick results to aid in immediate clinical decision-making.
CLIA Requirements: CLIA number and CLIA certification are required to perform the test.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.