Learn More
Electron Microscopy Sciences Holey Carbon Grid for Cryo TEM, Mesh: 200. Grid: Copper. Hole: 2.0 µm. Spacing: 2.0 µm., 10/PK
Supplier: Electron Microscopy Sciences CF222C10
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
C-flat is an ultra-flat, holey carbon-coated support grid for transmission electron microscopy (TEM). Unlike competing holey carbon films, C-flat is manufactured without plastics, so it is clean upon arrival and the user has no residue to contend with. Made with patent pending technology, C-flat provides an ultra-flat surface that results in better particle dispersion and more uniform ice thickness. Patterning is done using deep-UV projection lithography, ensuring the most accurate and consistent hole shapes and sizes down to submicron features. The precise methods by which C-flat is manufactured elminate artifacts such as excess carbon and edges around holes. C-flat holey carbon grids provide the ideal specimen support to achieve high resolution data in making them an ideal choice for cryo single particle analysis, cryo tomography and automated TEM analysis. Mesh: 200. Grid: Copper . Hole: 2.0 µm . Spacing: 2.0 µm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.