Learn More
Sterlitech Corporation CF042 Shim Assortment Pack (Stainless Steel): Includes 1ea (2 mil, 3 mil, 4 mil, 5 mil); 3ea (10 mil, 15mil)
Supplier: Sterlitech Corporation 1160030
CF042 Shims (Stainless Steel). Size: 9.207 x 4.572 cm for use in CF042.
Assortment pack, mix and match to make the optimal thickness. (a) 1 of 2 mil (0.051 mm); (b) 1 of 3 mil (0.076 mm); (c) 1 of 4 mil (0.102 mm); (d) 1 of 5 mil (0.127mm); (e) 3 of 10 mil (0.254mm); (f) 3 of 15 mil (0.381mm);