Learn More
Abnova™ CENYq12 CISH Probe
CENYq12 CISH Probe is designed for the qualitative detection of human chromosome Yq12 classical catellites III in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
Supplier: Abnova™ CC0003
Specifications
The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use. | |
The probe is intended to be used in combination with the CISH Implementation Kit 1, which provides necessary reagents for specimen pretreatment and post-hybridization processing. | |
Reagent Provided: 1. Digoxigenin-labeled polynucleotides targeting sequences mapping in Yq12 specific for the classical satellite III region DYZ1 of chromosome Y 2. Formamide based hybridization buffer |