Learn More
Abnova™ CEN6 (Green) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Supplier:  Abnova™ FC0185
Catalog No. 89-939-196
Add to Cart
Description
Description
Specifications
Specifications
Store at -20°C in the dark.
FISH
200 μL
Product Suggestions
Product Suggestions