Learn More
Alkali Scientific CellPro™ Aspirating Pipet, 2ML, Supplied without graduations or plugs, Individually Wrapped, Sterile, 800/cs
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Alkali Scientific SP202ACS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Serological Pipets • STERILE • Individually Wrapped Serological Pipettes • color coded paper/plastic wrapping for pop-through or peel-apart techniques • Stretched, Non-welded Design • Bi-directional and extra Negative Graduation Marks • Filter Plug and 100% Polystyrene • Gamma Sterilized; Pyrogen/Cytotoxin/RNase & Dnase Free