Learn More
Edvotek™ Cell Types in the Brain Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
The brain is an incredibly complex organ that uses a network of cells working together to function and perform everyday activities. This experiment helps students to examine the differences between cell types in the brain based on their profiles of proteins.
Supplier: Edvotek™ 1110
Description
- Storage: Some components require freezer storage
- For 6 Groups Sharing 3 Polyacrylamide Gels
Specifications
Kit | |
14 to 20 | |
12% Denaturing Polyacrylamide Gels (3), Vertical Gel Electrophoresis Apparatus, Power Supply, Shaker Platform (optional), Automatic Pipettes with Tips, Glacial Acetic Acid, Methanol, Beakers, Transfer Pipettes, Foil, Trays/Containers, Spatula, Lab Gloves, Safety Goggles, and Distilled Water | |
6 Lab Groups |
9 to Undergraduate | |
Pre-Stained Protein Standard Markers (Lyophilized), Protein Samples (Lyophilized), 10 x Tris-Glycine-SDS Chamber Buffer, Protein InstaStain™, Practice Gel Loading Solution, Transfer Pipettes | |
Some Components Require Freezer Storage |