Learn More
Cole-Parmer Argos Technologies Cell Strainer, 70 um, White, Sterile, Individual Wrap, 50/Cs
Supplier: Cole-Parmer 0439601
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Pair these cell strainers directly with your 50-mL centrifuge tubes. Choose from 40, 70 or 100 m mesh diameters. Their color-coded frames help differentiate sizes, while its extended tab makes handling easy. They are ready-to-use and each strainer is individually packaged and gamma sterilized.