Learn More
OriGene CEACAM5 Human shRNA Lentiviral Particle (Locus ID 1048)
SDP
Supplier:  OriGene TL314004V
Encompass_Preferred
CEACAM5 - Human shRNA lentiviral particles (4 unique 29mer target-specific shRNA, 1 scramble control), 0.5 ml each, >10^7 TU/ml.
Catalog No. NC2683885
Explore available promotions