Learn More
ACROBiosystems Human CD38&Serum Albumin Protein, His Tag (MALS verified)
Supplier: ACROBiosystems CDSH52H7100UG
Human CD38&Serum Albumin, His Tag (CDS-H52H7) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (CD38) & Asp 25 - Leu 609 (Serum Albumin) (Accession # P28907-1 (CD38) & P02768-1 (Serum Albumin)).