Learn More
TARGETMOL CHEMICALS INC CASEIN KINASE 1DELTA-IN- 5MG
Supplier: TARGETMOL CHEMICALS INC T60295
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Casein kinase 1(delta)-IN-1 (WAY-643895) is an inhibitor of casein kinase 1(delta) (CK1(delta)) which inhibits CK1(delta). Casein kinase 1(delta)-IN-1 can be used in the study of neurodegenerative diseases such as Alzheimer's disease. purity: 99%