Learn More
Hach Company Carrying Case for Sludge Judge Water Core Samplers
Supplier: Hach Company 2194301
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sludge Judge accessories help maintain a long, trouble-free service life. Cleaning brush not recommended for cleaning oil and grease. Use Grease Trap Cleaning Rod in conjunction with Cleaning Strips and a mild detergent to effectively remove oil & grease from plastic tubing.