Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Desktop Garden
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Enjoy a garden at your desk.
Supplier: Carolina Biological Supply 163865
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes:
- 3 gal. plastic round terrarium
- Gravel
- Soil
- Korean Rock Fern
- Vegetative Moss
- Fruticose Lichen
- Selaginella
- Instructions
Description
- 9 in.dia × 13 in.H (22.5 × 33cm)
- Assembly required
Specifications
For garden at desk | |
Ea. | |
3 gal. plastic round terrarium comes with all the materials for garden design, gravel, soil, korean rock fern, vegetative moss, Fruticose lichen and Selaginella, Size: 9 dia. x 13 H in., instructions | |
Desktop Garden |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction