Learn More
Thermo Scientific Dionex™ CarboPac™ PA1 IC Column
Supplier: Thermo Scientific 035391
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Analytical column, PEEK, 4mm x 250mm, maximum pressure: 4000 psi (275 bar), for ICS-2000, ICS-3000 IC Systems
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)