Learn More
Ted Pella Inc Carbon Type-B, 300 mesh, Copper, 50/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ted Pella Inc 01813F
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
A Formvar film coated with a “heavier” layer of carbon. This is the strongest and most versatile support film we produce. It is stable in the EM under all operating conditions including high magnification with high beam intensity. The films can withstand vigorous specimen preparation techniques. If the carbon surface is hydrophobic, specimen suspensions can be applied to the Formvar surface.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.