Learn More
Electron Microscopy Sciences Carbon Adhesive Tabs, 12 mm
Supplier: Electron Microscopy Sciences 7782512
Two sides of thick conductive adhesive (45 um) with the center conductive core film (35 um). Use for SEM including GSR. 125 um thick. No gas out-gassing. Traces of Si, Sb, S, Fe, Mg, Na.
Diameter: 12 mm