Learn More
Pyramex™ Capstone™ Shield - Safety Tear Off Visors
Catalog Number 19156096
Click to view available options
ForUseWithEquipment: Capstone™ Shield
Pack of 6 clear disposable, tear-off visors, suitable for high dust or muddy working environments
  • Fully adjustable elastic band
  • Fits over prescription glasses
  • Lens is coated to prevent fogging
  • Soft vinyl goggle body conforms to facial contours
  • Goggle provides protection against dust and chemical splash
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Removable vent caps allow the user to remove when more ventilation is required or leave in place for unsurpassed chemical splash resistance
  • Goggle incorporated with removable shield provides full face protection
  • 4 ventilation holes provide appropriate airflow
  • Ratchet shield allows user to open or close in 3 positions for adjustment
  • Polycarbonate shield conforms to the face offering increased impact and dust protection
  • For Use With (Equipment) Capstone™ Shield
    For Use With (Application) Suitable for high dust or muddy working environments
    Includes 6 clear disposable, tear-off visors