Learn More
Pyramex™ Capstone™ Shield
Click to view available options
Lens Tint:
Clear
IR/UV 3.0, Green
IR/UV 5.0, Green
Description
Specifications
Lens Type | H2X |
Certifications/Compliance | ANSI Z87.1 |
Description | Direct/Indirect Goggle with IR5 |
Lens Tint | IR/UV 5.0, Green |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |