Learn More
Perkin Elmer US LLC Caps for Disposable Micro Cells - Blue, Qty. 100
Supplier: Perkin Elmer US LLC L7110264
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Blue Caps for Disposable Micro Cells provide a tight closure, allowing samples to be stored in temperatures as low as -20°C. Also availabe in green and orange. 100 per package. Disposable micro cells sold separately.