Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-Sala™ ExoFit™ Full Body Harnesses
With quick connect and parachute buckles
$608.00 - $696.00
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-161-812
|
3M™
1108606 |
X-Large |
Each for $696.00
|
|
|||||
19-165-276
|
3M™
1108607 |
X-Small |
Each for $608.00
|
|
|||||
19-165-277
|
3M™
1108600 |
Small |
Each for $696.00
|
|
|||||
19-161-808
|
3M™
1108601 |
Medium | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-165-278
|
3M™
1108608 |
2X-Large | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Used as component of personal fall arrest system (arresting force must not exceed 1800 lbs. (8kN), for work positioning system to support user at work position, for climbing, rescue, controlled descent application and restraint system
- Designed for use by persons with combined weight (clothing, tools etc.) of no more than 352 lb. (160kg)
- Personal free fall system must be rigged to limit free fall to 6 ft., 2 ft. (work positioning) and 18 in. (climbing)
- Connectors (hooks, carabiners, and D-rings) must be capable of supporting at least 5000 lb. (22.2kN)
- X-shape wih wrap around no-tangle design
- Breathable dri-lex lining with polyester webbing
- Ergonomically designed shoulder, hip and leg padding
- With chest, shoulder and leg straps
- Fire resistant hook and loop fasteners
Positioning of D-Rings
- D-ring or attachment element is present on back, between shoulder blades
- Side D-rings for positioning or restraint applications only
- Front D-ring is for ladder climbing and not used positioning
- For rescue, back or front D-rings may be used
- D-rings on seat sling are for work positioning or personnel riding
Compliance
Meets CSA Z259.10, ANSI Z359.1, and OSHA requirements
Specifications
Back, Front, Side | |
5.5 lb. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title