Learn More
Cole-Parmer Torbal FSA5 Digital Cap Torque Tester, 44 lb in capacity
Supplier: Cole-Parmer 2530474
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Cap tester offers superior performance for the most demanding production or quality control applications. Engineered with high output loadcells the FSA TORQUE delivers accurate and repeatable torque measurements. With six modes and applications you can test any container or jar ranging from 0.79" (20 mm) to 7.09" (180 mm) in diameter. Addjustable steel posts with a non-slip rubber sleeve assures a firm grip on a wide range of container shapes and sizes. A die cast metal housing provides a stable platform for the torgue gauge and overall durability for a long life in demanding environments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.