Learn More
Cole-Parmer Fluke 718 30US Pressure Calibrator, 30 psig
Supplier: Cole-Parmer 6845089
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Fluke 718 Pressure Calibrator provides a total pressure calibration solution for transmitters, gauges and switches. This compact pressure calibration solution is about a third of the size of comparable instruments and weighs just one kilogram (two pounds). The unique easy to clean pump helps protect the pump from damage and provides the ability to service the pump without disassembly, even in the field.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.