Learn More
Cole-Parmer Fluke 718 100US Pressure Calibrator, 100 psig
Supplier:  Cole-Parmer 6845087
Encompass_Preferred
The Fluke 718 Pressure Calibrator provides a total pressure calibration solution for transmitters, gauges and switches. This compact pressure calibration solution is about a third of the size of comparable instruments and weighs just one kilogram (two pounds). The unique easy to clean pump helps protect the pump from damage and provides the ability to service the pump without disassembly, even in the field.
Catalog No. 50-993-412
Explore available promotions