Learn More
Cole-Parmer Fluke 717 100G Pressure Calibrator, 100 psig
Supplier: Cole-Parmer 6845079
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Pressure source and milliamp measurement capability to calibrate and maintain almost any pressure device with one tool
Error % calculation for quicker pass / fail decision making in the field
Best in class mA accuracy (0.015% for better e asuremetn confidence and workload coverage