Learn More
Electron Microscopy Sciences Calibration Standard Model 70-1D with Cert Mounted on Steel
Supplier: Electron Microscopy Sciences 801271DCAFM
A precision pattern providing accurate calibration in the horizontal plane for very high resolution, nanometer-scale measurements. Period: 70 nm pitch, one-dimensional array. Accurate to +/- 0.25 nm. Refer to calibration certificate for actual pitch. Surface: Silicon Oxide ridges on Silicon, 4x3 mm die. Ridge height (about 35 nm) and width (about 35 nm) are not calibrated. For AFM, use in contact, intermittent contact (TappingMode) and other modes with image sizes from 100 to 3000 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.