Learn More
Electron Microscopy Sciences Calibration Specimen 700-1D, Steel disk for AFM
Supplier: Electron Microscopy Sciences 801221DAFM
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Parallel Ridges Pitch 700, W-coated Photoresist on Si. Use contact or TappingMode.
Mount: AFM
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.