Learn More
Electron Microscopy Sciences Calibration Specimen 300-2D, Choose a mount
Supplier: Electron Microscopy Sciences 801232DX
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Array of Posts. Pitch 297. Al bumps on Si. Use contact or TappingMode.
Mount: Choose a mount
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.