Learn More
Crescent Chemical Co Inc CACODYLIC ACID-NA-SALT-3H2O
Supplier: Crescent Chemical Co Inc SE15540.02
Cacodylic acid-Na-salt-3H2O research grade For buffers, especially for bactericidal buffers used in electron microscopy. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 15540.02 CAS 6131-99-3