Learn More
Grainger CAB VAP ABSOR CRTRDG ACID FLAM
Supplier: Grainger 3NPJ2
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The 3M Cartridge, Acid Gas/Organic Vapor, 60926 is a respirator cartridge designed for use with 3M 6000, 7000 and FF-400 series respirator facepieces. It provides protection against certain organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen sulfide, and hydrogen fluoride. The cartridges use 3M's Advanced Electrostatic Media that is pleated for increased surface area. They have a low breathing resistance and high vapor capacity. The cartridges have a shelf life of 3 years from date of manufacture if unopened. They meet NIOSH approval requirements. The cartridges are sold individually and have an approximate weight of 0.35 lbs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.