Learn More
TARGETMOL CHEMICALS INC C16 CERAMIDE 25MG
Supplier: TARGETMOL CHEMICALS INC T37562
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. C16 Ceramide (Palmitoyl Ceramide) is a natural small molecule activating p53 through the direct and selective binding. purity: 99%