Learn More
Ace Glass, Inc. Buret, auto, 25mL, 24/40, 1:5 PTFE plug, complete with reservoir and pressure bulb/release tube, 1/10mL subdivision
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 573544
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)