Learn More
Bullard™ Frigitron™ 2000 Cool Tube Airline Respirator Accessory
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Accessory for use with Bullard ADP20 and EDP30 Free-Air™ Pumps.
Supplier: Bullard™ FRIGITRON2000
Description
Helps maintain worker comfort and maximize productivity while working in hot, humid environments
- Cools air by as much as 20°F (11°C)
- Features two adjustable airflow control valves that allow the user to adjust the incoming air to a temperature best suited to individual comfort
- NIOSH approved for use with Bullard's 88,CC20, GR50 or MB30 Series airline respirators
- 1/2 in. industrial interchange steel (Hansen compatible) quick-disconnect nipple
Specifications
FRIGITRON2000 | |
Cooling Assembly |