Learn More
Bullard™ 41 Series Breathing AirLine Filter Replacement Parts
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Replacement parts for use with 41 Series breathing airline filters.
$98.00
Specifications
For Use With (Equipment) | 41 Series Airline Filter |
---|
Alert:
The 41 Series Airline filters do not remove carbon monoxide and other toxic gases. Filter cartridge must be changed periodically for maximum efficiency. Frequency of cartridge changes depends on operating conditions. Cartridge should be changed immediately if respirator wearer feels, smells or tastes contaminants inside the respirator.
Catalog Number | Mfr. No. | Product Type | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Model | Price | Quantity & Availability | ||||
19-021409
![]() |
Bullard™
41AF |
Filter Cartridge | 41AF |
Each for $98.00
|
|
||||
19-824-401
![]() |
Bullard™
41P2M |
Manifold Assembly | 41P2M | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-824-403
![]() |
Bullard™
41P6UPK |
Manifold Assembly | 41P6UPK | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Replacement parts for 41 Series airlines
- Filter cartridge
- 6-outlet manifold upgrade for 41P6 includes:
- Manifold
- Regulator assembly
- Relief valve
- Gauge
- 2-outlet manifold assembly for 41P2 and 41P2E includes:
- Manifold with regulator
- Relief valve
- Gauge
Specifications
41 Series Airline Filter |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.