Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution for Nitrogen (Nitrate), Certified, LabChem™
$117.00 - $178.00
Specifications
Color | Clear to slightly turbid |
---|---|
pH Range | 3.0 ± 0.5 |
Format | Liquid |
Packaging | Amber Glass |
Storage Requirements | Room Temperature |
Specifications
Clear to slightly turbid | |
Liquid | |
Room Temperature |
3.0 ± 0.5 | |
Amber Glass | |
Buffer Solution for Nitrogen (Nitrate) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title