Learn More
TARGETMOL CHEMICALS INC BRIMONIDINE TARTRATE 500MG
Supplier: TARGETMOL CHEMICALS INC T6422
Also available in 100 mg and bulk. Please contact Fisher for quotes. Brimonidine Tartrate (AGN190342 tartrate) is a quinoxaline derivative and adrenergic (alpha)-2 receptor agonist (EC50 0.45 nM) that is used to manage intraocular pressure associated with open-angle glaucoma or ocular hypertension. purity: 99%