Learn More
TARGETMOL CHEMICALS INC BREFONALOL HCL 5MG
Supplier: TARGETMOL CHEMICALS INC T30586L
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Brefonalol HCl is an ADRA antagonist used in the treatment of neurological disorders and cardiovascular diseases and can be used in the study of angina pectoris arrhythmias and hypertension.