Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
Supplier: BRAND™ 701371
Description
- Easily apply to plates and remove without the use of expensive equipment.
- Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
- Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
- Polyethylene
- Polyester
- Aluminum
- Rayon
- Polypropylene
- Vinyl
Specifications
Vinyl | |
50 Sheets | |
Storage, fluorescence measurements | |
Sealing Film |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction