Learn More
Electron Microscopy Sciences Cryogenic Grid Box with Pin Type Lid for FEI Vitrobot, White/Blue
Supplier: Electron Microscopy Sciences 7116640
These specimen grid boxes are tools for storing or transferring cryogenic TEM specimen grids. Features four diamond shaped slots. Non-static clear covershield held in place with stainless steel screw, which is tapped in the center of the box. Box fits the FEI Vitrobot™, Gatan 626™, Gatan 3500™.
Color: White/Blue
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'