Learn More
List Biological Laboratories Inc Botulinum Neurotoxin Type B Light Chain, Recombinant, 10 μg
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: List Biological Laboratories Inc 620A
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development projects! Clostridium botulinum neurotoxins causes botulism in man and animals. Seven immunologically distinct serotypes of neurotoxin, designated types A through G, have been identified. Each is synthesized as a single polypeptide chain (MW 150,000), the holotoxin. When botulinum neurotoxin is exposed to proteases specific peptide bonds are cleaved or “nicked” resulting in the formation of a dichain molecule. Dichain neurotoxin is composed of a light chain region (MW 50,000) linked by a single disulfide bond and non-covalent interactions to a heavy chain (MW 100,000). The light chains are non-toxic proteins that retain the enzymatic activity encoded by the holotoxin. The Type B light chain cleaves the eukaryotic substrate synaptobrevin-2 and VAMPtide fluorogenic peptide substrates offered from List Labs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.