Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAV 100ML ADJ
Supplier: Wilmad Labglass LG3540110
Bottle, Specific Gravity, Gay-Lussac, Adjusted. Capacity: 100mL; Joint Size: [ST] 10/18. Adjusted at 20degreeC. The capillary stoppers are not interchangeable since the final adjustment for volume is made on the stopper.