Learn More
Wilmad Labglass BOTTLE SPC GRAV LE CHAT 300ML
Supplier: Wilmad Labglass LG3543100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Bottle, Specific Gravity, Le Chatelier, Class A. Used to determine the specific gravity of sand, cement and other fine materials.