Learn More
Wilmad Labglass BOTTLE SPC GRAV HUBBARD 24ML
Supplier: Wilmad Labglass LG3541100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Bottle, Specific Gravity, Hubbard. Capacity: 24mL. Used in standard test methods for specific gravity of asphalt cements, road tars, road oils and soft tar pitches.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov