Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton glass serum bottle, 100 mL, 144 case
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z114006144EA
100 mL volume, clear, O.D. 20 mm, diam. X H 52 mm x 95 mm. It is autoclavable, easy sterilization, is used to contain substances to prevent contamination.