Learn More
Enzo Life Sciences Bordetella pertussis FHA (100 µg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences ALX6301230100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alternative name: Filamentous hemagglutinin antigen. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized from 0.05M TRIS-HCl and 0.5M sodium chloride at pH 8, 5% sucrose. Appearance: White powder. Source: From Bordetella pertussis, strain NCTC 10739. UniProt: P12255. Application: ELISA, LINE assay, Antibody capture. Long Term Storage: +4°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.